N-[(4-Methylphenyl)sulphonyl]acetamide ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI EUROPE
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

TCIAM3123-250MGEA 1590 DKK
TCIAM3123-250MG
N-[(4-Methylphenyl)sulphonyl]acetamide ≥98.0% (by HPLC, titration analysis)
N-[(4-Methylphenyl)sulphonyl]acetamide
Formel: C₉H₁₁NO₃S
Molekylvægt: 213,25 g/mol
MDL Number: MFCD00159373
CAS-nummer: 1888-33-1

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 138.0 to 142.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR