Moclobemide ≥98.0% (by HPLC)

Leverandør: TCI EUROPE
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

TCIAM2733-10MGEA 854 DKK
TCIAM2733-10MG
Moclobemide ≥98.0% (by HPLC)
Moclobemide
Formel: C₁₃H₁₇ClN₂O₂
Molekylvægt: 268,74 g/mol
MDL Number: MFCD00865388
CAS-nummer: 71320-77-9

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Elemental analysis (Nitrogen) 9.90 to 10.90 %
Melting point 136.0 to 140.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR