2-Methylisonicotinic acid ≥98.0% (by GC, titration analysis)

Leverandør: TCI EUROPE
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

TCIAM2602-1GEA 1100 DKK
TCIAM2602-1G
2-Methylisonicotinic acid ≥98.0% (by GC, titration analysis)
2-Methylisonicotinic acid
Formel: C₇H₇NO₂
Molekylvægt: 137,14 g/mol
MDL Number: MFCD00513435
CAS-nummer: 4021-11-8

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR