Methyl pyrimidine-2-carboxylate ≥98.0% (by GC)

Leverandør: TCI EUROPE
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

TCIAM2456-1GEA 731 DKK
TCIAM2456-1G
Methyl pyrimidine-2-carboxylate ≥98.0% (by GC)
Methyl pyrimidine-2-carboxylate
Formel: C₆H₆N₂O₂
Molekylvægt: 138,1200 g/mol
MDL Number: MFCD03426929
CAS-nummer: 34253-03-7

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 102.0 to 106.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR