1-(Methylsulphonyl)piperazine ≥98.0% (by GC)
Leverandør: TCI EUROPE
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
TCIAM2437-5GEA
600
DKK
TCIAM2437-5G
1-(Methylsulphonyl)piperazine ≥98.0% (by GC)
1-(Methylsulphonyl)piperazine
Formel:
C₅H₁₂N₂O₂S Molekylvægt: 164,23 g/mol |
MDL Number:
MFCD02153225 CAS-nummer: 55276-43-2 |
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 97.0 % |
Melting point | 94.0 to 98.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...