4-Methylthiazole-5-carboxylic acid ≥97.0% (by GC, titration analysis)
Leverandør: TCI EUROPE
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
TCIAM2235-5GEA
526
DKK
TCIAM2235-5G
4-Methylthiazole-5-carboxylic acid ≥97.0% (by GC, titration analysis)
4-Methylthiazole-5-carboxylic acid
Formel:
C₅H₅NO₂S Molekylvægt: 143,17 g/mol Smeltepunkt: 260°C |
MDL Number:
MFCD00626872 CAS-nummer: 20485-41-0 |
Specifikationer Testresultater
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 97.0 % |
Purity (Neutralization titration) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...