5-Methoxy-2-nitrophenylamine ≥98.0% (by GC)

Leverandør: TCI EUROPE
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

TCIAM2054-5GEA 747 DKK
TCIAM2054-5G
5-Methoxy-2-nitrophenylamine ≥98.0% (by GC)
5-Methoxy-2-nitrophenylamine
Formel: C7H8N2O3
Molekylvægt: 168,15 g/mol
MDL Number: MFCD00179573
CAS-nummer: 16133-49-6
EINECS: 240-293-4

Bestil nu

Specifikationer Testresultater

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (GC) min. 98.0 %
Melting point 130.0 to 134.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR