Lanthanum(III) isopropoxide

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

TCIAL0174-200MGEA 206 DKK
TCIAL0174-200MG
Lanthanum(III) isopropoxide
Lanthanum(III) isopropoxide
Formel: C₉H₂₁LaO₃
Molekylvægt: 316,18
MDL Number: MFCD00015305
CAS-nummer: 19446-52-7
UN nummer: 1325
ADR: 4.1,III

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow powder to crystal
Content of Lanthanum 41.5 to 46.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR