2-Iodo-5-phenylthiophene ≥97.0% (by GC)

Leverandør: TCI EUROPE
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

TCIAI0998-1GEA 928 DKK
TCIAI0998-1G
2-Iodo-5-phenylthiophene ≥97.0% (by GC)
2-Iodo-5-phenylthiophene
Formel: C₁₀H₇IS
Molekylvægt: 286,13 g/mol
MDL Number: MFCD13180547
CAS-nummer: 13781-37-8

Bestil nu

Specifikationer Testresultater

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 97.0 %
Melting point 82.0 to 86.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR