Imidazo[1,2-a]pyridine ≥98.0% (by GC)

Leverandør: TCI EUROPE
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

TCIAI0922-5GEA 501 DKK
TCIAI0922-5G
Imidazo[1,2-a]pyridine ≥98.0% (by GC)
Imidazo[1,2-a]pyridine
Formel: C₇H₆N₂
Kogepunkt: 98°C
MDL Number: MFCD00005553
CAS-nummer: 274-76-0

Bestil nu

Specifikationer Testresultater

Appearance Light yellow to Amber to Dark green clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR