Thiazol-2-ylmethanol ≥98.0% (by GC)

Leverandør: TCI EUROPE
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

TCIAH1476-1GEA 780 DKK
TCIAH1476-1G
Thiazol-2-ylmethanol ≥98.0% (by GC)
Thiazol-2-ylmethanol
Formel: C₄H₅NOS
Molekylvægt: 115,1500 g/mol
Kogepunkt: 76°C
MDL Number: MFCD06200855
CAS-nummer: 14542-12-2

Bestil nu

Specifikationer Testresultater

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 62.0 to 68.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR