Homophthalic anhydride ≥98.0% (by GC)
Leverandør: TCI EUROPE
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Advarsel
TCIAH1346-5GEA 386 DKK
TCIAH1346-5G TCIAH1346-25G
Homophthalic anhydride ≥98.0% (by GC)
Homophthalic anhydride
Formel: C₉H₆O₃
Molekylvægt: 162,14 g/mol
Kogepunkt: 161°C
MDL Number: MFCD00006894
CAS-nummer: 703-59-3
EINECS: 211-873-4
Bestil nu
Specifikationer Testresultater
Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Morpholine Method) min. 96.0 %