Hydrogeniodid 57%
Leverandør: TCI EUROPE
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
TCIAH1221-300MLEA
780
DKK
TCIAH1221-300ML
Hydrogeniodid 57%
Hydrogeniodid
Formel:
HI Molekylvægt: 127,91 g/mol Kogepunkt: 127°C |
MDL Number:
MFCD00011347 CAS-nummer: 10034-85-2 EINECS: 233-109-9 UN nummer: 1787 ADR: 8,II |
Specifikationer Testresultater
Appearance | Colorless to Red to Green clear liquid |
Concentration (Neutralization titration) | min. 57.0 w/w% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...