Glycidyl acrylate ≥95.0% (by GC) stabilised

Leverandør: TCI EUROPE
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig


Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

TCIAG0497-5GEA 936 DKK
TCIAG0497-5G
Glycidyl acrylate ≥95.0% (by GC) stabilised
Glycidyl acrylate
Formel: C₆H₈O₃
Molekylvægt: 128,13 g/mol
Kogepunkt: 65°C
Flammepunkt: 61°C
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00005138
CAS-nummer: 106-90-1
EINECS: 203-440-3
UN nummer: 2929
ADR: 6.1,II

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR