2-Fluoro-3-formylphenylboronic acid (contains varying amounts of Anhydride)

Leverandør: TCI EUROPE
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

TCIAF1078-1GEA 444 DKK
TCIAF1078-1G
2-Fluoro-3-formylphenylboronic acid (contains varying amounts of Anhydride)
2-Fluoro-3-formylphenylboronic acid
Formel: C₇H₆BFO₃
Molekylvægt: 167,9300 g/mol
MDL Number: MFCD07369725
CAS-nummer: 849061-98-9

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 to 112.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR