2,2-Difluoro-2-(fluorosulphonyl)acetic acid ≥95.0% (by titrimetric analysis)

Leverandør: TCI EUROPE
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

TCIAF0500-5GEA 353 DKK
TCIAF0500-5G
2,2-Difluoro-2-(fluorosulphonyl)acetic acid ≥95.0% (by titrimetric analysis)
2,2-Difluoro-2-(fluorosulphonyl)acetic acid
Formel: C₂HF₃O₄S
Kogepunkt: 153°C
Massefylde: 1,72
Opbevaringstemperatur: Køleskab
MDL Number: MFCD01320807
CAS-nummer: 1717-59-5
UN nummer: 3265
ADR: 8,II

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (Neutralization back titration) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR