4-(Ethoxycarbonylmethyl)phenylboronic acid pinacol ester ≥98.0% (by titrimetric analysis)

Leverandør: TCI EUROPE
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

TCIAE1393-1GEA 1040 DKK
TCIAE1393-1G
4-(Ethoxycarbonylmethyl)phenylboronic acid pinacol ester ≥98.0% (by titrimetric analysis)
4-(Ethoxycarbonylmethyl)phenylboronic acid pinacol ester
Formel: C₁₆H₂₃BO₄
Molekylvægt: 290,1624 g/mol
MDL Number: MFCD08704684
CAS-nummer: 859169-20-3

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 95.0 %
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR