4-(Ethoxycarbonyl)-2-nitrophenylboronic acid ≥98.0% (by HPLC) (contains varying amounts of Anhydride)

Leverandør: TCI EUROPE
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

TCIAE1203-1GEA 1040 DKK
TCIAE1203-1G
4-(Ethoxycarbonyl)-2-nitrophenylboronic acid ≥98.0% (by HPLC) (contains varying amounts of Anhydride)
4-(Ethoxycarbonyl)-2-nitrophenylboronic acid
Formel: C₉H₁₀BNO₆
Molekylvægt: 238,9898 g/mol
Smeltepunkt: 125°C
MDL Number: MFCD02179460
CAS-nummer: 5785-70-6

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) 97.0 to 108.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR