Ethyl-4-(1-hydroxy-1-methylethyl)-2-propylimidazole-5-carboxylate ≥96.0% (by GC, titration analysis)
Leverandør: TCI EUROPE
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
TCIAE0976-25GEA
838
DKK
TCIAE0976-25G
Ethyl-4-(1-hydroxy-1-methylethyl)-2-propylimidazole-5-carboxylate ≥96.0% (by GC, titration analysis)
Ethyl-4-(1-hydroxy-1-methylethyl)-2-propylimidazole-5-carboxylate
Formel:
C₁₂H₂₀N₂O₃ Molekylvægt: 240,2988 g/mol |
MDL Number:
MFCD07787565 CAS-nummer: 144689-93-0 |
Specifikationer Testresultater
Appearance | Light orange to Yellow to Green powder to crystal |
Purity (GC) | min. 96.0 % |
Purity (Nonaqueous Titration) | min. 96.0 % |
Melting point | 102.0 to 106.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...