2,4-Dimethyl-1,3-dioxolane cis- and trans-mixture ≥98.0% (by GC)

Leverandør: TCI EUROPE
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

TCIAD5748-25GEA 419 DKK
TCIAD5748-25G
2,4-Dimethyl-1,3-dioxolane cis- and trans-mixture ≥98.0% (by GC)
2,4-Dimethyl-1,3-dioxolane
Formel: C₅H₁₀O₂
Molekylvægt: 102,13 g/mol
Kogepunkt: 91°C
MDL Number: MFCD00190653
CAS-nummer: 3390-12-3
EINECS: 222-221-3
UN nummer: 1993
ADR: 3,II

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 % (total of isomers)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR