5,6-Difluoro-4,7-diiodo-2,1,3-benzothiadiazole ≥98.0% (by GC)

Leverandør: TCI EUROPE
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

TCIAD5547-200MGEA 1720 DKK
TCIAD5547-200MG
5,6-Difluoro-4,7-diiodo-2,1,3-benzothiadiazole ≥98.0% (by GC)
5,6-Difluoro-4,7-diiodo-2,1,3-benzothiadiazole
Formel: C₆F₂I₂N₂S
Molekylvægt: 423,95 g/mol
CAS-nummer: 1293389-29-3

Bestil nu

Specifikationer Testresultater

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 205.0 to 209.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR