5,6-Difluoro-2,1,3-benzothiadiazole ≥98.0% (by GC)

Leverandør: TCI EUROPE
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

TCIAD5246-1GEA 1740 DKK
TCIAD5246-1G
5,6-Difluoro-2,1,3-benzothiadiazole ≥98.0% (by GC)
5,6-Difluoro-2,1,3-benzothiadiazole
Formel: C₆H₂F₂N₂S
Molekylvægt: 172,15 g/mol
MDL Number: MFCD28100486
CAS-nummer: 1293389-28-2

Bestil nu

Specifikationer Testresultater

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 66.0 to 70.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR