(2-((Dimethylamino)methyl)phenyl)boronic acid (contains varying amounts of Anhydride)

Leverandør: TCI EUROPE
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

TCIAD5165-1GEA 1150 DKK
TCIAD5165-1G
(2-((Dimethylamino)methyl)phenyl)boronic acid (contains varying amounts of Anhydride)
(2-((Dimethylamino)methyl)phenyl)boronic acid
Formel: C₉H₁₄BNO₂
Molekylvægt: 179,0240 g/mol
MDL Number: MFCD01318999
CAS-nummer: 85107-53-5

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) 98.0 to 111.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR