BD140 ≥98.0% (by GC) for albumin binding assay

Leverandør: TCI EUROPE
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

TCIAD4898-25MGEA 1440 DKK
TCIAD4898-25MG TCIAD4898-100MG
BD140 ≥98.0% (by GC) for albumin binding assay
BD140
Formel: C₂₁H₂₁BF₂N₂O
Molekylvægt: 366,22 g/mol
CAS-nummer: 1201643-08-4

Bestil nu

Specifikationer Testresultater

Purity (GC) min. 98.0 %
Melting point 153.0 to 157.0 deg-C
Lambda max. 563.0 to 567.0 nm (CH2Cl2)
Molar extinction coefficient min. 10000 (CH2Cl2, 563.0 to 567.0nm)
Human serum albmin (HSA) binding activity S/N ratio min. 10

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR