2,4-Dichloro-5-nitropyridine ≥98.0% (by GC)

Leverandør: TCI EUROPE
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

TCIAD4752-1GEA 772 DKK
TCIAD4752-1G
2,4-Dichloro-5-nitropyridine ≥98.0% (by GC)
2,4-Dichloro-5-nitropyridine
Formel: C₅H₂Cl₂N₂O₂
Molekylvægt: 192,9900 g/mol
MDL Number: MFCD07368834
CAS-nummer: 4487-56-3

Bestil nu

Specifikationer Testresultater

Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 42.0 to 46.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR