N-(2,6-Dimethylphenyl)-2-(piperazin-1-yl)acetamide ≥98.0% (by GC, titration analysis)

Leverandør: TCI EUROPE
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

TCIAD4319-25GEA 706 DKK
TCIAD4319-25G
N-(2,6-Dimethylphenyl)-2-(piperazin-1-yl)acetamide ≥98.0% (by GC, titration analysis)
N-(2,6-Dimethylphenyl)-2-(piperazin-1-yl)acetamide
Formel: C₁₄H₂₁N₃O
Molekylvægt: 247,34 g/mol
MDL Number: MFCD06384973
CAS-nummer: 5294-61-1

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 116.0 to 120.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR