Dihydrodaidzein ≥93.0% (by HPLC)

Leverandør: TCI EUROPE
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

TCIAD4239-25MGEA 1790 DKK
TCIAD4239-25MG
Dihydrodaidzein ≥93.0% (by HPLC)
Dihydrodaidzein
Formel: C₁₅H₁₂O₄
Molekylvægt: 256,26 g/mol
Opbevaringstemperatur: Fryser
MDL Number: MFCD00210494
CAS-nummer: 17238-05-0

Bestil nu

Specifikationer Testresultater

Appearance Light orange to Yellow to Green powder to crystal
Purity (HPLC) min. 93.0 area%
Melting point 244.0 to 250.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR