2,4-Diaminoanisole sulphate hydrate ≥98.0% (by HPLC)

Leverandør: TCI EUROPE
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

TCIAD1309-25GEA 805 DKK
TCIAD1309-25G
2,4-Diaminoanisole sulphate hydrate ≥98.0% (by HPLC)
2,4-Diaminoanisole sulphate hydrate
Formel: C₇H₁₂N₂O₅S·H₂O
MDL Number: MFCD00012978
CAS-nummer: 6219-67-6
EINECS: 228-290-6

Bestil nu

Specifikationer Testresultater

Appearance White to Brown to Dark purple powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 96.0% (calcd.on anh.substance)
Solubility in hot Water within almost transparency
Drying loss max. 20.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR