2,5-Dichlorobenzenesulphonic acid ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI EUROPE
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

TCIAD0334-25GEA 952 DKK
TCIAD0334-25G
2,5-Dichlorobenzenesulphonic acid ≥98.0% (by HPLC, titration analysis)
2,5-Dichlorobenzenesulphonic acid
Formel: C₆H₄Cl₂O₃S
MDL Number: MFCD00035751
CAS-nummer: 88-42-6
EINECS: 201-829-2
UN nummer: 3261
ADR: 8,III

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 95.0 %
Melting point 97.0 to 101.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR