4'-Chloro-2'-nitroacetophenone ≥98.0% (by GC)
Leverandør: TCI EUROPE
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
TCIAC3308-1GEA
846
DKK
TCIAC3308-1G
4'-Chloro-2'-nitroacetophenone ≥98.0% (by GC)
4'-Chloro-2'-nitroacetophenone
Formel:
C₈H₆ClNO₃ Molekylvægt: 199,5900 g/mol Kogepunkt: 157°C |
MDL Number:
MFCD09031331 CAS-nummer: 23082-51-1 |
Specifikationer Testresultater
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 55.0 to 59.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...