4-Chloro-2-formylphenylboronic acid (contains varying amounts of Anhydride)

Leverandør: TCI EUROPE
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

TCIAC3223-1GEA 796 DKK
TCIAC3223-1G
4-Chloro-2-formylphenylboronic acid (contains varying amounts of Anhydride)
4-Chloro-2-formylphenylboronic acid
Formel: C₇H₆BClO₃
Molekylvægt: 184,39 g/mol
MDL Number: MFCD08689492
CAS-nummer: 913835-76-4

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 to 111.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR