4-Chloro-2-iodophenol ≥98.0% (by GC, titration analysis)
Leverandør: TCI EUROPE
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
TCIAC3172-1GEA
600
DKK
TCIAC3172-1G
4-Chloro-2-iodophenol ≥98.0% (by GC, titration analysis)
4-Chloro-2-iodophenol
Formel:
C₆H₄ClIO Molekylvægt: 254,4500 g/mol |
MDL Number:
MFCD08235248 CAS-nummer: 71643-66-8 |
Specifikationer Testresultater
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 75.0 to 79.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...