5-Chloro-1-methyl-4-nitroimidazole ≥98.0% (by total nitrogen basis)

Leverandør: TCI EUROPE
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

TCIAC1646-5GEA 649 DKK
TCIAC1646-5G
5-Chloro-1-methyl-4-nitroimidazole ≥98.0% (by total nitrogen basis)
5-Chloro-1-methyl-4-nitroimidazole
Formel: C₄H₄ClN₃O₂
Molekylvægt: 161,55 g/mol
CAS-nummer: 4897-25-0
EINECS: 225-521-2

Bestil nu

Specifikationer Testresultater

Appearance White to Orange to Green powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Melting point 145.0 to 150.0 deg-C
Solubility in hot Methanol very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR