Sodium hexanoate ≥99.0% (by titrimetric analysis)
Leverandør: TCI EUROPE
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
TCIAC1187-25GEA
1380
DKK
TCIAC1187-25G
Sodium hexanoate ≥99.0% (by titrimetric analysis)
Sodium hexanoate
Formel:
C₆H₁₁NaO₂ Molekylvægt: 138,1401 g/mol |
MDL Number:
MFCD00059056 CAS-nummer: 10051-44-2 EINECS: 233-179-0 |
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (Nonaqueous Titration) | min. 99.0 % (after drying) |
Drying loss | max. 3.0 % |
Sulfate | to pass test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...