(Bromoethynyl)triisopropylsilane ≥95.0% (by GC, titration analysis)

Leverandør: TCI EUROPE
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

TCIAB5412-1GEA 993 DKK
TCIAB5412-1G
(Bromoethynyl)triisopropylsilane ≥95.0% (by GC, titration analysis)
(Bromoethynyl)triisopropylsilane
Formel: C₁₁H₂₁BrSi
Molekylvægt: 261,28 g/mol
MDL Number: MFCD11036302
CAS-nummer: 111409-79-1

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Red to Green clear liquid
Purity (GC) min. 95.0 %
Purity (Argentmetric Titration) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR