1,2-Dibromo-2,4-dicyanobutane ≥98.0% (by GC)

Leverandør: TCI EUROPE
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

TCIAB5387-5GEA 222 DKK
TCIAB5387-5G TCIAB5387-25G
1,2-Dibromo-2,4-dicyanobutane ≥98.0% (by GC)
1,2-Dibromo-2,4-dicyanobutane
Formel: C₆H₆Br₂N₂
Kogepunkt: 212°C
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00072483
CAS-nummer: 35691-65-7
EINECS: 252-681-0
UN nummer: 3077
ADR: 9,III

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 53.0 to 58.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR