7-Bromo-2-(chloroacetyl)-9,9-difluorofluorene ≥98.0% (by GC)

Leverandør: TCI EUROPE
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

TCIAB5130-1GEA 764 DKK
TCIAB5130-1G
7-Bromo-2-(chloroacetyl)-9,9-difluorofluorene ≥98.0% (by GC)
7-Bromo-2-(chloroacetyl)-9,9-difluorofluorene
Formel: C₁₅H₈BrClF₂O
Molekylvægt: 357,58 g/mol
MDL Number: MFCD28400046
CAS-nummer: 1378387-81-5

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 183.0 to 187.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR