Bis(3-bromophenyl)acetylene ≥97.0% (by GC)

Leverandør: TCI EUROPE
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

TCIAB4853-1GEA 961 DKK
TCIAB4853-1G
Bis(3-bromophenyl)acetylene ≥97.0% (by GC)
Bis(3-bromophenyl)acetylene
Formel: C₁₄H₈Br₂
Molekylvægt: 336,03 g/mol
MDL Number: MFCD11111154
CAS-nummer: 153404-60-5

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow powder to crystal
Purity (GC) min. 97.0 %
Melting point 104.0 to 108.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR