4,7-Bis(5-bromo-2-thienyl)-2-n-octyl-2H-benzotriazole ≥90.0% (by GC)

Leverandør: TCI EUROPE
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

TCIAB4542-100MGEA 1150 DKK
TCIAB4542-100MG
4,7-Bis(5-bromo-2-thienyl)-2-n-octyl-2H-benzotriazole ≥90.0% (by GC)
4,7-Bis(5-bromo-2-thienyl)-2-n-octyl-2H-benzotriazole
Formel: C₂₂H₂₃Br₂N₃S₂
Molekylvægt: 553,38 g/mol
CAS-nummer: 1254062-41-3

Bestil nu

Specifikationer Testresultater

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (GC) min. 90.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR