6-Bromoquinoxaline ≥98.0% (by GC)
Leverandør: TCI EUROPE
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
TCIAB4476-5GEA
1240
DKK
TCIAB4476-5G
TCIAB4476-1G
6-Bromoquinoxaline ≥98.0% (by GC)
6-Bromoquinoxaline
Formel:
C₈H₅BrN₂ Molekylvægt: 209,0427 g/mol Kogepunkt: 149°C |
MDL Number:
MFCD00837757 CAS-nummer: 50998-17-9 |
Specifikationer Testresultater
Appearance | Light yellow to Yellow to Orange powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 96.0 % |
Melting point | 54.0 to 58.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...