4-Bromo-1-naphthaleneboronic acid (contains varying amounts of Anhydride)

Leverandør: TCI EUROPE
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

TCIAB4463-1GEA 485 DKK
TCIAB4463-1G
4-Bromo-1-naphthaleneboronic acid (contains varying amounts of Anhydride)
4-Bromo-1-naphthaleneboronic acid
Formel: C₁₀H₈BBrO₂
Molekylvægt: 250,88
MDL Number: MFCD08701764
CAS-nummer: 145965-14-6

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 to 108.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR