[5,15-Bis(phenylethynyl)-10,20-bis[(triisopropylsilyl)ethynyl]porphyrinato]magnesium(II) ≥95.0% (by HPLC)

Leverandør: TCI EUROPE
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

TCIAB4314-50MGEA 3530 DKK
TCIAB4314-50MG
[5,15-Bis(phenylethynyl)-10,20-bis[(triisopropylsilyl)ethynyl]porphyrinato]magnesium(II) ≥95.0% (by HPLC)
[5,15-Bis(phenylethynyl)-10,20-bis[(triisopropylsilyl)ethynyl]porphyrinato]magnesium(II)
Formel: C₅₈H₆₀MgN₄Si₂
Molekylvægt: 893,62 g/mol
CAS-nummer: 1397288-30-0

Bestil nu

Specifikationer Testresultater

Purity (HPLC) min. 95.0 area%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR