1-Bromo-3-hexylbenzene ≥98.0% (by GC)
Leverandør: TCI EUROPE
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
TCIAB4180-5GEA
526
DKK
TCIAB4180-5G
1-Bromo-3-hexylbenzene ≥98.0% (by GC)
1-Bromo-3-hexylbenzene
Formel:
C₁₂H₁₇Br Kogepunkt: 98°C |
MDL Number:
MFCD09909567 CAS-nummer: 38409-59-5 |
Specifikationer Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...