1-Boc-azetidine-3-ylmethanol ≥98.0% (by GC)

Leverandør: TCI EUROPE
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

TCIAB3992-1GEA 1040 DKK
TCIAB3992-1G
1-Boc-azetidine-3-ylmethanol ≥98.0% (by GC)
1-Boc-azetidine-3-ylmethanol
Formel: C₉H₁₇NO₃
Molekylvægt: 187,24 g/mol
MDL Number: MFCD06656141
CAS-nummer: 142253-56-3

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to lump
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR