1-Benzoylpiperazine ≥98.0% (by GC, titration analysis)
Leverandør: TCI EUROPE
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
TCIAB3676-5GEA
468
DKK
TCIAB3676-5G
1-Benzoylpiperazine ≥98.0% (by GC, titration analysis)
1-Benzoylpiperazine
Formel:
C₁₁H₁₄N₂O Molekylvægt: 190,24 g/mol Kogepunkt: 207°C |
MDL Number:
MFCD00810192 CAS-nummer: 13754-38-6 |
Specifikationer Testresultater
Appearance | White to Almost white powder to crystaline |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...