L-Glutamic acid 5-benzyl ester ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI EUROPE
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

TCIAB3316-5GEA 362 DKK
TCIAB3316-5G
L-Glutamic acid 5-benzyl ester ≥98.0% (by HPLC, titration analysis)
L-Glutamic acid 5-benzyl ester
Formel: C₁₂H₁₅NO₄
Molekylvægt: 237,3 g/mol
MDL Number: MFCD00002633
CAS-nummer: 1676-73-9
EINECS: 216-826-1

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 157.0 to 162.0 deg-C
Specific rotation [a]20/D +17.0 to +21.0 deg (C=1、AcOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR