Benzamidoxime ≥98.0% (by HPLC, titration analysis)
Leverandør: TCI EUROPE
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
TCIAB0015-25GEA
1470
DKK
TCIAB0015-25G
Benzamidoxime ≥98.0% (by HPLC, titration analysis)
Benzamidoxime
Formel:
C₇H₈N₂O Molekylvægt: 136,15 g/mol |
MDL Number:
MFCD00031485 CAS-nummer: 613-92-3 EINECS: 210-361-8 |
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 76.0 to 78.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...