Napabucasin ≥98.0% (by GC)

Leverandør: TCI EUROPE
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

TCIAA3110-100MGEA 1150 DKK
TCIAA3110-100MG TCIAA3110-25MG
Napabucasin ≥98.0% (by GC)
Napabucasin
Formel: C₁₄H₈O₄
Molekylvægt: 240,21 g/mol
Opbevaringstemperatur: Fryser
MDL Number: MFCD28155270
CAS-nummer: 83280-65-3

Bestil nu

Specifikationer Testresultater

Appearance Light yellow to Amber to Dark green powder to crystaline
Purity (GC) min. 98.0 %
Melting point 225.0 to 229.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR