6-Amino-5-bromoquinoxaline ≥98.0% (by GC, titration analysis)
Leverandør: TCI EUROPE
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
TCIAA2913-25GEA
3020
DKK
TCIAA2913-25G
TCIAA2913-5G
6-Amino-5-bromoquinoxaline ≥98.0% (by GC, titration analysis)
6-Amino-5-bromoquinoxaline
Formel:
C₈H₆BrN₃ Molekylvægt: 224,0573 g/mol |
MDL Number:
MFCD00799024 CAS-nummer: 50358-63-9 |
Specifikationer Testresultater
Appearance | Light yellow to Amber to Dark green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 155.0 to 159.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...