4-Amino-3-iodobenzoic acid ≥98.0% (by titrimetric analysis)

Leverandør: TCI EUROPE
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

TCIAA2873-1GEA 961 DKK
TCIAA2873-1G
4-Amino-3-iodobenzoic acid ≥98.0% (by titrimetric analysis)
4-Amino-3-iodobenzoic acid
Formel: C₇H₆INO₂
Molekylvægt: 263,03 g/mol
MDL Number: MFCD01660550
CAS-nummer: 2122-63-6

Bestil nu

Specifikationer Testresultater

Purity (HPLC) min. 97.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 206.0 to 210.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR